Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scarf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004157.2 | NP_001004157.2 | 11 | 2124 | ||
GenBank mRNA | AK133477.1 | - | 11 | 2107 | ||
AK220215.1 | - | 11 | 2108 | |||
BC057600.1 | - | 10 | 2031 | |||
BC080288.1 | - | 9 | 1855 | 6411836 |