Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP5-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004325.1 | NP_001004325.1 | 1 | 1101 | ||
GenBank mRNA | AJ628245.1 | - | 1 | 1101 |
Gene Symbol: KRTAP5-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005405.2 | NP_001005405.1 | 1 | 1004 | ||
GenBank mRNA | AJ628244.1 | - | 1 | 426 |