Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2A25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004488.1 | NP_001004488.1 | 1 | 298 | ||
GenBank mRNA | BC136814.1 | - | 1 | 336 | 9020804 | |
BC136829.1 | - | 1 | 336 | 9020820 |