Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005274.1 | NP_001005274.1 | 1 | 107 | ||
GenBank mRNA | BC140748.1 | - | 1 | 157 | 9021616 |