Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005895.2 | NP_001005895.2 | 7 | 711 | ||
GenBank mRNA | AY751756.1 | - | 7 | 653 |