Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JMJD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005920.2 | NP_001005920.2 | 9 | 872 | ||
NM_001323918.1 | NP_001310847.1 | 9 | 874 | |||
NM_001323919.1 | NP_001310848.1 | 8 | 744 | |||
NM_001323920.1 | NP_001310849.1 | 8 | 789 | |||
NM_001323922.1 | NP_001310851.1 | 8 | 784 | |||
NR_136650.1 | 8 | 962 | ||||
NR_136651.1 | 8 | 967 | ||||
NR_136652.1 | 7 | 877 | ||||
GenBank mRNA | AF370420.1 | - | 9 | 756 | ||
AL524336.3 | - | 8 | 847 | |||
AL524922.3 | - | 8 | 854 | |||
BC073785.1 | - | 8 | 844 | |||
BC094850.1 | - | 6 | 490 | |||
BC137100.1 | - | 9 | 838 | 9021097 | ||
BC137101.1 | - | 9 | 838 | 9021098 | ||
BG284477.1 | - | 4 | 329 | |||
BG423721.1 | - | 5 | 384 | |||
BG826580.1 | - | 6 | 670 | |||
BX335015.2 | - | 9 | 753 | |||
DQ074695.1 | - | 8 | 662 |