Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008811.1 | NP_001008811.1 | 4 | 716 | ||
XM_006242351.2 | XP_006242413.1 | 4 | 772 | |||
GenBank mRNA | AB355640.1 | - | 4 | 716 |