Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017811.3 | NP_060281.2 | 1, 2 | 759 | ||
XM_011517949.1 | XP_011516251.1 | 1, 2 | 779 | |||
GenBank mRNA | AJ240087.1 | - | 1, 2 | 190 | ||
AK000426.1 | - | 1, 2 | 376 | |||
AK225880.1 | - | 1, 2 | 364 | |||
BC004862.2 | - | 1, 2 | 366 | 3838157 | ||
BC047584.1 | - | 1, 2 | 615 | 4827644 | ||
CR457233.1 | - | 1, 2 | 185 |