Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnal4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009666.1 | NP_001009666.1 | 4 | 1384 | ||
GenBank mRNA | BC087688.1 | - | 4 | 1384 | 7311257 |
Gene Symbol: RGD1559781 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256369.2 | XP_006256431.1 | 1 | 1198 |