Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oas2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009715.1 | NP_001009715.1 | 9 | 1992 | ||
GenBank mRNA | AY230746.1 | - | 9 | 1992 |