Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD36C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001310154.1 | NP_001297083.1 | 82, 83 | 5608 | ||
GenBank mRNA | AK123626.1 | - | 29, 30 | 2456 | ||
AL832836.1 | - | 41, 42 | 2268 | |||
U28831.1 | - | 16, 17 | 1112 |