Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klrb1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010964.1 | NP_001010964.1 | 6 | 869 | ||
GenBank mRNA | FQ232553.1 | - | 5 | 734 | ||
M62891.1 | - | 6 | 869 |