Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFIT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010987.2 | NP_001010987.1 | 2 | 811 | ||
GenBank mRNA | BC137368.1 | - | 2 | 781 | 9021366 | |
BC137369.1 | - | 2 | 781 | 9021367 | ||
BC150189.1 | - | 2 | 793 | 40148511 | ||
BX647300.1 | - | 2 | 811 |