Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011700.2 | NP_001011700.2 | 2 | 243 | ||
GenBank mRNA | BC132994.1 | - | 2 | 225 | 40147016 | |
BC132996.1 | - | 2 | 225 | 40147018 |