Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sf3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011986.1 | NP_001011986.1 | 2 | 197 | ||
XM_006240885.2 | XP_006240947.1 | 1 | 154 | |||
XM_006240886.2 | XP_006240948.1 | 2 | 451 | |||
GenBank mRNA | BC079320.1 | - | 2 | 197 | 7135238 |