Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cadm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012201.1 | NP_001012201.1 | 10 | 1304 | ||
XM_006242946.2 | XP_006243008.1 | 9 | 1213 | |||
GenBank mRNA | BC078966.1 | - | 10 | 1304 | 7111523 |