Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhgb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012215.1 | NP_001012215.1 | 2 | 3974 | ||
GenBank mRNA | BC087084.1 | - | 2 | 3974 | 7189313 |
Gene Symbol: Pcdhga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037140.1 | NP_001032217.1 | 3 | 3803 | ||
GenBank mRNA | AY574008.1 | - | 3 | 3803 |
Gene Symbol: Pcdhga10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037135.1 | NP_001032212.1 | 2 | 3944 | ||
GenBank mRNA | AY574009.1 | - | 2 | 3944 |
Gene Symbol: Pcdhga11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037153.1 | NP_001032230.1 | 2 | 3943 | ||
GenBank mRNA | AY574010.1 | - | 2 | 3943 |
Gene Symbol: Pcdhga12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037337.1 | NP_001032414.1 | 2 | 3993 | ||
GenBank mRNA | AY574011.1 | - | 2 | 3993 |
Gene Symbol: Pcdhga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037139.1 | NP_001032216.1 | 3 | 3973 | ||
GenBank mRNA | AY574012.1 | - | 3 | 3973 |
Gene Symbol: Pcdhga3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037154.1 | NP_001032231.1 | 3 | 3967 | ||
GenBank mRNA | AY574014.1 | - | 3 | 3967 |
Gene Symbol: Pcdhga5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037137.1 | NP_001032214.1 | 3 | 3974 | ||
GenBank mRNA | AY574016.1 | - | 3 | 3974 |
Gene Symbol: Pcdhga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014773.1 | NP_001014773.1 | 3 | 4016 | ||
GenBank mRNA | AY574018.1 | - | 3 | 4016 |
Gene Symbol: Pcdhga8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037156.1 | NP_001032233.1 | 3 | 3944 | ||
GenBank mRNA | AY574019.1 | - | 3 | 3944 |
Gene Symbol: Pcdhga9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037158.1 | NP_001032235.1 | 3 | 3987 | ||
GenBank mRNA | AY574020.1 | - | 3 | 3987 |
Gene Symbol: Pcdhgb8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037159.1 | NP_001032236.1 | 2 | 3969 | ||
GenBank mRNA | AY574027.1 | - | 2 | 3969 |
Gene Symbol: Pcdhgc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053943.1 | NP_446395.1 | 3 | 3869 | ||
GenBank mRNA | AY574028.1 | - | 3 | 3869 |