Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mup20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012323.1 | NP_001012323.1 | 7 | 769 | ||
GenBank mRNA | BC089613.1 | - | 7 | 769 | 6746470 | |
BC092096.1 | - | 7 | 795 | 4161847 |