Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012324.2 | NP_001012324.1 | 6 | 1427 | ||
GenBank mRNA | AK038812.1 | - | 6 | 1289 | ||
AK040995.1 | - | 6 | 1451 | |||
AK134746.1 | - | 6 | 1381 | |||
AK163501.1 | - | 5 | 3449 | |||
BC065151.1 | - | 6 | 1297 | 5708030 | ||
BC089559.1 | - | 6 | 1427 | 30295705 |