Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOHLH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012415.2 | NP_001012415.2 | 4 | 456 | ||
NM_001101677.1 | NP_001095147.1 | 4 | 456 | |||
XM_005263403.3 | XP_005263460.1 | 4 | 407 | |||
XM_006717109.3 | XP_006717172.1 | 5 | 1532 | |||
XM_011518698.2 | XP_011517000.1 | 6 | 1561 | |||
GenBank mRNA | AY884306.1 | - | 4 | 401 | ||
AY902244.1 | - | 4 | 456 | |||
BC031861.1 | - | 4 | 467 | 4826967 | ||
EB710265.1 | - | 2 | 153 |