Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRTAP5-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012503.1 | NP_001012521.1 | 1 | 581 | ||
GenBank mRNA | AB126076.1 | - | 1 | 581 | ||
AJ628243.1 | - | 1 | 167 | |||
BC136967.1 | - | 1 | 570 | 9020959 | ||
BC136968.1 | - | 1 | 570 | 9020960 |