Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PQLC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040125.1 | NP_001035214.1 | 8 | 1667 | ||
NM_001040126.1 | NP_001035215.1 | 8 | 1055 | |||
NM_001287531.1 | NP_001274460.1 | 8 | 1024 | |||
NM_017765.2 | NP_060235.2 | 9 | 1297 | |||
NR_109848.1 | 9 | 1297 | ||||
XM_005245916.2 | XP_005245973.1 | 8 | 1080 | |||
XM_006710710.2 | XP_006710773.1 | 9 | 1190 | |||
XM_006710711.2 | XP_006710774.1 | 8 | 1049 | |||
XM_011541634.2 | XP_011539936.1 | 8 | 2064 | |||
GenBank mRNA | AK000327.1 | - | 9 | 1218 | ||
AK125580.1 | - | 8 | 1585 | |||
AK130171.1 | - | 9 | 1215 | |||
AK130175.1 | - | 9 | 1217 | |||
AK172745.1 | - | 8 | 942 | |||
BC015324.1 | - | 8 | 960 | 4426038 |