Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013196.1 | NP_001013214.1 | 7 | 925 | ||
XM_006230859.2 | XP_006230921.1 | 7 | 917 | |||
XM_006230860.2 | XP_006230922.1 | 7 | 882 | |||
XM_006230861.2 | XP_006230923.1 | 6 | 1080 | |||
XM_006230862.2 | XP_006230924.1 | 6 | 1100 | |||
XM_008760159.1 | XP_008758381.1 | 7 | 919 | |||
GenBank mRNA | BC088201.1 | - | 7 | 925 | 7378025 |