Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017768.4 | NP_060238.3 | 2 | 338 | ||
XM_005271013.1 | XP_005271070.1 | 2 | 239 | |||
XM_017001695.1 | XP_016857184.1 | 2 | 210 | |||
GenBank mRNA | AK022951.1 | - | 2 | 252 | ||
BC003407.1 | - | 2 | 138 | |||
BC008586.1 | - | 2 | 236 | 2900193 | ||
DA310461.1 | - | 2 | 339 |