siRNA Details
Gene Symbol: Rabl2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013221.1 | NP_001013239.1 | 9 | 1121 | ||
XM_006242225.2 | XP_006242287.1 | 9 | 1205 | |||
XM_006242226.2 | XP_006242288.1 | 9 | 1203 | |||
XM_006242227.1 | XP_006242289.1 | 9 | 926 | |||
XM_006242228.2 | XP_006242290.1 | 9 | 1161 | |||
GenBank mRNA | BC083624.1 | - | 9 | 1121 | 7134121 |

More Information
Other Species Targeted

