Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pea15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013231.1 | NP_001013249.1 | 4 | 1695 | ||
GenBank mRNA | BC085766.1 | - | 4 | 1695 | 7190459 | |
FQ235065.1 | - | 1 | 810 |