Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnase1l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013232.1 | NP_001013250.1 | 1 | 213 | ||
XM_006251893.2 | XP_006251955.1 | 2 | 344 |