Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013434.1 | NP_001013452.1 | 6 | 2144 | ||
GenBank mRNA | BC091333.1 | - | 6 | 2144 | 7314509 |