Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIGNL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013690.4 | NP_001013712.4 | 2 | 2990 | ||
XM_005268870.3 | XP_005268927.1 | 2 | 3043 | |||
XM_017019298.1 | XP_016874787.1 | 2 | 3232 | |||
XM_017019299.1 | XP_016874788.1 | 3 | 3788 | |||
GenBank mRNA | AK129980.1 | - | 4 | 756 |