Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014158.1 | NP_001014180.1 | 6 | 2416 | ||
XM_006228003.2 | XP_006228065.1 | 5 | 2149 | |||
XM_006228004.2 | XP_006228066.1 | 5 | 2529 | |||
XM_006228006.2 | XP_006228068.1 | 5 | 2612 | |||
XM_006228007.2 | XP_006228069.1 | 4 | 5008 | |||
XM_008758764.1 | XP_008756986.1 | 5 | 2369 | |||
XM_008758765.1 | XP_008756987.1 | 5 | 3960 | |||
GenBank mRNA | BC085827.1 | - | 6 | 2416 | 7131994 |