Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014166.1 | NP_001014188.1 | 8 | 824 | ||
XM_008760344.1 | XP_008758566.1 | 8 | 862 | |||
GenBank mRNA | BC081993.1 | - | 8 | 824 | 7130307 | |
FQ212643.1 | - | 8 | 852 |