Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poll | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014168.1 | NP_001014190.1 | 3 | 239 | ||
XM_006231545.2 | XP_006231607.1 | 2 | 145 | |||
GenBank mRNA | BC085841.1 | - | 3 | 239 | 7135812 |