Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014204.1 | NP_001014226.1 | 2 | 873 | ||
XM_006257363.2 | XP_006257425.1 | 2 | 1135 | |||
GenBank mRNA | BC086949.1 | - | 2 | 873 | 7325189 |