Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naa60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014226.2 | NP_001014248.2 | 2 | 177 | ||
XM_008767519.1 | XP_008765741.1 | 1 | 75 | |||
XM_008767520.1 | XP_008765742.1 | 2 | 213 | |||
GenBank mRNA | AY316589.1 | - | 2 | 219 | ||
BC105304.1 | - | 2 | 177 | 7191187 | ||
FQ212054.1 | - | 2 | 181 |