Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glod4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014227.1 | NP_001014249.1 | 9 | 926 | ||
GenBank mRNA | BC088458.1 | - | 9 | 926 | 7127323 | |
FQ210234.1 | - | 9 | 944 | |||
FQ230415.1 | - | 9 | 902 |