Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024901.1 | NP_001020072.1 | 2 | 544 | ||
XM_008773422.1 | XP_008771644.1 | 2 | 646 | |||
GenBank mRNA | DQ058659.1 | - | 2 | 544 |