Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dram2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025018.1 | NP_001020189.1 | 7 | 797 | ||
XM_006233120.2 | XP_006233182.1 | 7 | 741 | |||
XM_006233121.2 | XP_006233183.1 | 6 | 681 | |||
XM_006233122.2 | XP_006233184.1 | 6 | 782 | |||
XM_006233123.2 | XP_006233185.1 | 7 | 743 | |||
GenBank mRNA | BC091251.1 | - | 7 | 797 | 7372838 |
Gene Symbol: LOC689412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109538.1 | NP_001103008.1 | 1 | 444 |