Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025019.1 | NP_001020190.1 | 4 | 811 | ||
XM_006233919.2 | XP_006233981.1 | 5 | 1636 | |||
XM_008761670.1 | XP_008759892.1 | 3 | 701 | |||
GenBank mRNA | BC091294.1 | - | 4 | 811 | 7307721 | |
FQ216821.1 | - | 4 | 826 |