Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdyhv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025024.1 | NP_001020195.1 | 6 | 1286 | ||
GenBank mRNA | BC091306.1 | - | 6 | 1286 | 7311328 |