Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161407.1 | NP_001154879.1 | 8 | 2158 | ||
NM_019080.2 | NP_061953.2 | 8 | 2218 | |||
XM_006719840.3 | XP_006719903.1 | 8 | 2617 | |||
GenBank mRNA | AB032991.1 | - | 8 | 1982 | ||
AB097019.1 | - | 8 | 2190 | |||
AB097028.1 | - | 8 | 2190 | |||
AB097029.1 | - | 8 | 2190 | |||
AB097030.1 | - | 8 | 1868 | |||
AB097031.1 | - | 8 | 1868 | |||
AK001723.1 | - | 8 | 2011 |