Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPIRE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032451.1 | NP_115827.1 | 14 | 1874 | ||
XM_011523379.1 | XP_011521681.1 | 14 | 2640 | |||
XM_011523380.2 | XP_011521682.1 | 15 | 1896 | |||
XM_011523381.1 | XP_011521683.1 | 15 | 2720 | |||
XM_011523384.1 | XP_011521686.1 | 13 | 1870 | |||
XM_017023779.1 | XP_016879268.1 | 14 | 1886 | |||
XM_017023780.1 | XP_016879269.1 | 14 | 2051 | |||
XM_017023781.1 | XP_016879270.1 | 13 | 1523 | |||
GenBank mRNA | AB058735.1 | - | 12 | 1972 | ||
AJ422077.1 | - | 14 | 1864 | |||
AK094918.1 | - | 7 | 691 | |||
AK130685.1 | - | 4 | 1756 | |||
AL834408.1 | - | 14 | 1668 | |||
BC063706.1 | - | 6 | 487 | |||
BC139732.1 | - | 12 | 1492 | 40075864 |