Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2d12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201360.1 | NP_958748.1 | 2 | 299 | ||
GenBank mRNA | AK078880.1 | - | 2 | 299 | ||
AK165474.1 | - | 1173 | ||||
BC026417.1 | - | 3 | 197 | |||
BC089596.1 | - | 1 | 1173 | |||
BC139169.1 | - | 2 | 283 | 8862191 | ||
BC139170.1 | - | 2 | 283 | 8862192 | ||
BC145567.1 | - | 2 | 283 | 9054132 |