Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204856.1 | NP_001191785.1 | 5 | 557 | ||
NM_024041.3 | NP_076946.1 | 5 | 508 | |||
NR_037937.1 | 5 | 508 | ||||
GenBank mRNA | AK056322.1 | - | 5 | 477 | ||
AK301644.1 | - | 5 | 501 | |||
AV718162.2 | - | 5 | 434 | |||
BC000264.1 | - | 5 | 452 | 3356277 | ||
BU540926.1 | - | 5 | 374 |
Gene Symbol: TNFAIP8L2-SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204848.1 | NP_001191777.1 | 5 | 420 | ||
GenBank mRNA | BG531092.1 | - | 5 | 386 | ||
CR999387.1 | - | 4 | 401 |