Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270411.1 | NP_001257340.1 | 8 | 1258 | ||
NM_004900.4 | NP_004891.4 | 8 | 1333 | |||
GenBank mRNA | AK024854.1 | - | 7 | 1613 | ||
BC053859.1 | - | 7 | 1205 | 6174285 | ||
BX114955.1 | - | 3 | 371 | |||
U61083.1 | - | 5 | 781 | |||
U61084.1 | - | 8 | 1322 |
Gene Symbol: APOBEC3A_B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193289.1 | NP_001180218.1 | 5 | 899 |