Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRCH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032773.3 | NP_116162.1 | 18, 19 | 2041 | ||
XM_005269362.2 | XP_005269419.1 | 18, 19 | 2412 | |||
XM_005269363.2 | XP_005269420.1 | 17, 18 | 2340 | |||
XM_005269365.2 | XP_005269422.1 | 17, 18 | 2304 | |||
XM_005269367.2 | XP_005269424.1 | 16, 17 | 2232 | |||
XM_006713791.3 | XP_006713854.1 | 18, 19 | 2412 | |||
XM_006713792.3 | XP_006713855.1 | 17, 18 | 2340 | |||
XM_006713793.2 | XP_006713856.1 | 17, 18 | 2304 | |||
XM_017007350.1 | XP_016862839.1 | 18, 19 | 2412 | |||
XM_017007351.1 | XP_016862840.1 | 16, 17 | 2286 | |||
XM_017007352.1 | XP_016862841.1 | 15, 16 | 2178 | |||
GenBank mRNA | AK024647.1 | - | 3, 4 | 247 | ||
AK055819.1 | - | 13, 14 | 1147 | |||
AK293745.1 | - | 18, 19 | 2030 | |||
AK300466.1 | - | 15, 16 | 1541 | |||
AK303521.1 | - | 17, 18 | 1847 | |||
AL137527.1 | - | 1, 2 | 120 | |||
BC007504.2 | - | 18, 19 | 1993 | 2960711 | ||
BC010565.1 | - | 3, 4 | 266 |