Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024335.1 | NP_001019506.1 | 6 | 1130 | ||
XM_006237380.2 | XP_006237442.1 | 5 | 2861 | |||
XM_008763293.1 | XP_008761515.1 | 6 | 2656 | |||
GenBank mRNA | BC095844.1 | - | 6 | 1130 | 7389613 |