Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024360.2 | NP_001019531.1 | 6 | 1224 | ||
XM_008766878.1 | XP_008765100.1 | 5 | 2200 | |||
GenBank mRNA | AB210855.1 | - | 6 | 1124 | ||
BC128712.1 | - | 6 | 1224 | 7133504 |