Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025063.1 | NP_001020234.1 | 5 | 826 | ||
XM_006236512.2 | XP_006236574.1 | 5 | 814 | |||
XM_008762916.1 | XP_008761138.1 | 5 | 678 | |||
GenBank mRNA | BC079152.1 | - | 5 | 826 | 7128646 |