Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iqcf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025153.1 | NP_001020324.1 | 8 | 657 | ||
XM_006243837.1 | XP_006243899.1 | 8 | 693 | |||
GenBank mRNA | BC079282.1 | - | 8 | 657 | 7134209 |