Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198410.3 | NP_940802.1 | 8 | 981 | ||
GenBank mRNA | AK005086.1 | - | 2 | 220 | ||
AK139144.1 | - | 8 | 982 | |||
AY424295.1 | - | 8 | 982 | |||
BC030379.1 | - | 3 | 606 | |||
BC120512.1 | - | 7 | 944 | 40129435 | ||
BC125428.1 | - | 7 | 944 | 40129943 | ||
BC144954.1 | - | 6 | 1020 |
Set Membership: |
G-protein coupled receptor |